Learn More
R&D Systems™ H-Met-Gly-Pro-AMC Fluorogenic Peptide Substrate
Substrate for Methionine Aminopeptidase 1, 1D and 2
Brand:  R&D Systems™ ES017
Product Code. 18177990
  • 297.00 € / 5mg
Explore more special offers
Description
Description
  • Hydrolysis of Pro-AMC amide bond releases AMC, a highly fluorescent group
  • A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
  • Excellent substrate for methionine aminopeptidases; after the enzymatic removal of the Met residue, the substrate can be cleaved by DPPIV/CD26-like enzymes
  • Purity >97% by TLC
Specifications
Specifications
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.