Learn More
R&D Systems™ Recombinant Human Erythropoietin/EPO (Tissue Culture Grade)
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 287-TC-500
482.93 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18308767
  • 641.00 € / 1 set
Explore more special offers
Description
Description
The Recombinant Human Erythropoietin (Tissue Culture Grade) is derived from CHO. The Recombinant Human Erythropoietin (Tissue Culture Grade) has been validated for the following applications: Bioactivity.
Specifications
Specifications
NP_000790.2
2056
Erythropoietin/EPO
Chinese Hamster Ovary cell line, CHO-derived human Erythropoietin/EPO protein Ala28-Arg193
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Recombinant
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
21 kDa
500 U
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.