Learn More
R&D Systems™ Human FPRL1/FPR2 (NP_001453) VersaClone cDNA
Formyl Peptide Receptor-like 1
Brand:  R&D Systems™ RDC0059
Product Code. 18492310
  • 322.00 € / 10µg
Explore more special offers
Description
Description
The Human FPRL1/FPR2 (NP_001453) VersaClone cDNA clone contains the complete ORF for FPRL1/FPR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FPRL1/FPR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hFPRL1/FPR2
10 μg at 0.2 μg/μL
1068bp
Human
NP_001453
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human FPRL1/FPR2 (NP_001453) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans