Learn More
R&D Systems™ Human ICAM-5 (NP_003250) VersaClone cDNA
Intercellular Adhesion Molecule 5
Brand: R&D Systems™ RDC0824
This item is not returnable.
View return policy
Description
The Human ICAM-5 (NP_003250) VersaClone cDNA clone contains the complete ORF for ICAM-5, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ICAM-5 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hICAM5 | |
10 μg at 0.2 μg/μL | |
2788bp | |
Human |
NP_003250 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans