Learn More
R&D Systems™ Human MCHR1 (AAC14587) VersaClone cDNA
Melanin-concentrating Hormone Receptor 1
Brand: R&D Systems™ RDC0568
This item is not returnable.
View return policy
Description
The Human MCHR1 (AAC14587) VersaClone cDNA clone contains the complete ORF for MCHR1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MCHR1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hMCHR1 | |
10 μg at 0.2 μg/μL | |
1222bp | |
Human |
AAC14587 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans