missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Methionine Aminopeptidase 1 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 3537-ZN-010
416.40 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC (Catalog # ES017 ). The resulting GP-AMC is cleaved by Recombinant Human DPPIV/CD26 (Catalog # 9168-SE ). The specific activity is >200 pmol/min/μg, as measured under the described conditions. SSpecifications
P53582 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 38 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
METAP1 | |
Recombinant | |
Methionine Aminopeptidase 1/METAP1 |
Enzyme Activity | |
23173 | |
Methionine Aminopeptidase 1/METAP1 | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Methionine Aminopeptidase 1/METAP1 protein His52-Phe386 & Lys53-Phe386, with a C-terminal 10-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |