Learn More
R&D Systems™ Human Methylcellulose Serum-Free Enriched Media
Complete, defined media for the differentiation and enumeration of human hematopoietic stem cells
Brand:  R&D Systems™ HSC005SF
425.90 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18422190
  • 542.00 € / 100mL
Explore more special offers
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Methylcellulose Serum-Free Enriched Media
100 mL