Learn More
R&D Systems™ Human/Mouse/Rat Neural Progenitor Cell Marker Antibody Panel
For the identification and characterization of neural progenitor cells
Brand:  R&D Systems™ SC025
Product Code. 18462100
  • 947.00 € / 1 set
Explore more special offers
Includes: 25μg each of antibodies to Notch-1, CXCR4, Vimentin, SSEA-1, Musashi-1, SOX1, SOX2, and Nestin supplied as 10X stock
Description
Description
Includes eight antibodies to increase confidence in cell identification and characterization
Compatible with detection of SSEA-1 and CXCR4 by flow cytometry, which can be completed in less than two hours
Compatible with detection of all markers by immun
Specifications
Specifications
1 Kit
Store the unopened product at 2°C to 8°C. Do not use past expiration date.