Learn More
R&D Systems™ Human OATP1B3/SLCO1B3 (NP_062818) VersaClone cDNA
Solute Carrier Organic Anion Transporter Family, Member 1B3
Brand:  R&D Systems™ RDC0870
Product Code. 18739498
  • 747.00 € / 10µg
Explore more special offers
Description
Description
The Human OATP1B3/SLCO1B3 (NP_062818) VersaClone cDNA clone contains the complete ORF for OATP1B3/SLCO1B3/OATP8, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The OATP1B3/SLCO1B3/OATP8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hSLCO1B3
10 μg at 0.2 μg/μL
2121bp
Human
NP_062818
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans