Learn More
R&D Systems™ MagCellect Human B Cell Isolation Kit
Designed to isolate B cells via a negative selection principle
Brand:  R&D Systems™ MAGH103
Product Code. 18461690
  • 762.00 € / 1 set
Explore more special offers
Description
Description
The resulting cell preparation is highly enriched with B cells
Typical recovery of the targeted cell population ranges between 45% and 65%
Purity of recovered B cells ranges between 90 and 98%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.