Learn More
R&D Systems™ MagCellect Human CD8+ T Cell Isolation Kit
Designed to isolate CD8+ T cells via a negative selection principle
Brand:  R&D Systems™ MAGH112
Product Code. 18462600
  • 762.00 € / 1 set
Explore more special offers
Description
Description
CD8 is a heterodimeric glycoprotein consisting of an alpha and beta chain. It is expressed on cytolytic T cells and functions in conjunction with the T cell receptor in the recognition of MHC/peptide complexes.
Typical purity of recovered CD8+ T cells ranges between 75% and 87%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.