Learn More
R&D Systems™ MagCellect Human NK Cell Isolation Kit
Designed to isolate Natural Killer cells via a negative selection principle
Brand:  R&D Systems™ MAGH109
Product Code. 18472280
  • 1200.00 € / 1 set
Explore more special offers
Description
Description
Typical purity of recovered NK cells ranges between 80% and 90%
MagCellect Streptavidin Ferrofluid is added to the reaction which allows the streptavidin-coated nanoparticles to interact with biotinylated antibody tagged cells, the unwanted cell fraction
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.