Learn More
R&D Systems™ MagCellect Mouse B Cell Isolation Kit
Designed to isolate B cells via a negative selection principle
Brand:  R&D Systems™ MAGM204
Product Code. 18472480
  • 647.00 € / 1 set
Explore more special offers
Description
Description
Typical recovery ranges between 30% and 55% and the purity of recovered B cells ranges from 90-97%
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction), leaving the untagged cells or desired cell population in suspension to be harvested by aspiration while the tube remains in the magnetic field
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.