Learn More
R&D Systems™ MagCellect Mouse NK Cell Isolation Kit
Designed to isolate Natural Killer cells via a negative selection principle
Brand:  R&D Systems™ MAGM210
Product Code. 18401360
  • 647.00 € / 1 set
Explore more special offers
Description
Description
The purity of recovered cells ranges between 70% and 80%
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction), leaving the untagged cells or desired cell population in suspension to be harvested by aspiration while the tube remains in the magnetic field
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.