Learn More
R&D Systems™ MagCellect Rat B Cell Isolation Kit
Designed to isolate B cells via a negative selection principle
Brand:  R&D Systems™ MAGR303
Product Code. 18431270
  • 626.00 € / 1 set
Explore more special offers
Description
Description
Typical recovery of the targeted cell population ranges between 40% and 65% and the purity of recovered B cells ranges from 89-95%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
Magnetically tagged cells will migrate toward the magnet (unwanted cell fraction)
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.