missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mca-RPKPVE-Nval-WRK(Dnp)-NH2 Fluorogenic MMP Substrate

Applications: Bioactivity, Binding Activity

Brand:  R&D Systems™ ES002

256.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18186970

  • 336.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The peptide substrate contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2,4-dinitrophenyl group. It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence. It is an excellent substrate for MMP-3 (stromelysin 1) and MMP-10 (stromelysin 2). The cleavage site is the peptide bond between Glu and Nval (norvaline).

It is also an excellent substrate for Trypsin, HGF activator and factor Xa.

Specifications

Specifications

Liquid
Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 10.0mg/mL or 4.3 mM.
Purity: >95%, by HPLC, Source: Synthetic
Bioactivity, Binding Activity
Predicted MW: 1674 Da
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers