missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mca-SEVNLDAEFRK(Dnp)RR-NH2 Fluorogenic Peptide Substrate

Applications: Bioactivity

Brand:  R&D Systems™ ES004

256.58 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18196490

  • 336.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Purity 93.8% by HPLC
  • Contains the Swedish mutation of the amyloid precursor protein (APP) beta-secretase cleavage site
  • A K-R-R sequence is added for easy solubilization

Specifications

Specifications

Liquid
Supplied as a stock solution in dimethyl sulfoxide (DMSO) (100μL) at a concentration of 10.0mg/mL or 4.1 mM..
Source: Synthetic
Bioactivity
Predicted MW: 2001 Da
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers