Learn More
R&D Systems™ Mouse CCR7 (NP_031745) VersaClone cDNA
Chemokine (C-C motif) Receptor 7
Brand: R&D Systems™ RDC0015
This item is not returnable.
View return policy
Description
The Mouse CCR7 (NP_031745) VersaClone cDNA clone contains the complete ORF for CCR7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CCR7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mCCR7 | |
10 μg at 0.2 μg/μL | |
1149bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
NP_031745 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
Mouse |
For Reasearch Use Only; Not for Use in Humans