Learn More
R&D Systems™ Mouse Frizzled-8 (NP_032084) VersaClone cDNA
FZD8
Brand:  R&D Systems™ RDC0293
Product Code. 18402400
  • 747.00 € / 10µg
Explore more special offers
Description
Description
The Mouse Frizzled-8 (NP_032084) VersaClone cDNA clone contains the complete ORF for Frizzled-8, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Frizzled-8 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
mFZD8
10 μg at 0.2 μg/μL
2071bp
Mouse
NP_032084
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans