Learn More
R&D Systems™ Mouse NrCAM (NP_795904) VersaClone cDNA
Neuronal Cell Adhesion Molecule
Brand:  R&D Systems™ RDC0810
Product Code. 18402600
  • 1170.00 € / 10µg
Explore more special offers
Description
Description
The Mouse NrCAM (NP_795904) VersaClone cDNA clone contains the complete ORF for NrCAM, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The NrCAM insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
mNRCAM
10 μg at 0.2 μg/μL
3784bp
Mouse
NP_795904
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans