Learn More
R&D Systems™ N21-MAX Insulin Free Media Supplement (50X)
Tested in the viability and growth of E18 rat hippocampal neurons
Brand:  R&D Systems™ AR010
Product Code. 18716397
  • 137.00 € / 10mL
Explore more special offers
Includes: The following 20 components for long-term neuronal cell culture: Albumin (bovine); L-Carnitine; Catalase; Corticosterone; Ethanolamine; Glutathione; Galactose; Holo-Transferrin; Linoleic Acid; Linolenic Acid; Lipoic Acid; Progesterone; Putrescine; Retinyl acetate; Retinol; Selenite; Superoxide dismutase; Triiodo-L-thyronine; D,L-alpha-Tocopherol; D,L-alpha-Tocopherol acetate;
Description
Description
N21-MAX Insulin Free Media Supplement is supplied as a 50X concentrated solution. A serum-free media supplement specially optimized for the long-term culture of neurons.
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 mL