Learn More
R&D Systems™ Proteome Profiler Human Chemokine Array Kit
The Proteome Profiler Human Chemokine Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
Brand:  R&D Systems™ ARY017
Product Code. 18077631
  • 646.00 € / 1 set
Explore more special offers
Description
Description
The Proteome Profiler Human Chemokine Array Kit contains 4 membranes - each spotted in duplicate with 31 different chemokine antibodies.
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.