Learn More
R&D Systems™ Rat CD27/TNFRSF7 (NP_001019506) VersaClone cDNA
CD27
Brand: R&D Systems™ RDC0357
This item is not returnable.
View return policy
Description
The Rat CD27/TNFRSF7 (NP_001019506) VersaClone cDNA clone contains the complete ORF for CD27/TNFRSF7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD27/TNFRSF7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
CD27/TNFRSF7 | |
10 μg at 0.2 μg/μL | |
768bp | |
CD27, CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
Rat |
NP_001019506 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
939 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Product Content Correction
Product Title
R&D Systems™ Rat CD27/TNFRSF7 (NP_001019506) VersaClone cDNA > 10 μg
For Reasearch Use Only; Not for Use in Humans
Spot an opportunity for improvement?Share a Content Correction