Learn More
R&D Systems™ Recombinant Human MMP-12 Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
Brand:  R&D Systems™ WBC019
139.09 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18324128
  • 180.00 € / Each
Explore more special offers
Description
Description
The Recombinant Human MMP-12 Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifications
Specifications
Western Blot
MMP-12 Western Blot Standard
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Unconjugated
MMP-12
4321
1 Vial
MMP12
Recombinant
Product Suggestions