Learn More
R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X)
Optimized for the extended culture of pluripotent stem cell-derived cardiomyocytes. R&D Systems™ StemXVivo Cardiomyocyte Maintenance Media Supplement (50X) is nutrient-rich, fully defined, and serum-free.
Brand:  R&D Systems™ AR011
Product Code. 18766747
  • 173.00 € / 10mL
Explore more special offers
Alert:
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.
Description
Description
  • Maintains healthy cardiomyocyte population for longer than 200 days
  • High lot-to-lot consistency decreases experimental variation
Repeated mechanical stress can negatively affect cell attachment and survival. For extended culture, daily media exchanges should be handled as gently as possible.
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Liquid
Detection of Cardiac Troponin T in Human ES-derived Cardiomyocytes, Troponin I Expression in Human ES-derived Cardiomyocytes
RUO
50 X
Maintenance Media Supplement
10 mL