Learn More
R&D Systems™ StemXVivo Chondrogenic Base Media
Base media for the differentiation of MSCs into chondrocytes
Brand:  R&D Systems™ CCM005
161.55 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18472130
  • 206.00 € / 50mL
Explore more special offers
Description
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into chondrocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Offers flexibility to evaluate novel cytokine and growth factor combinations to induce chondrogenesis
  • Has been developed and optimized using MSCs
  • Can be used with StemXVivo Human/Mouse or Rat Chondrogenic Supplements to reduce variation during chondrogenesis
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Stem Cell Expansion Media
50 mL