Learn More
R&D Systems™ StemXVivo Osteogenic/Adipogenic Base Media
Base media for the differentiation of MSCs into osteocytes or adipocytes
Brand:  R&D Systems™ CCM007
Product Code. 18431550
  • 216.00 € / 250mL
Explore more special offers
Description
Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes or adipocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
  • Supports reproducible human, mouse, and rat MSC differentiation
  • Offers flexibility to evaluate novel cytokine and growth factor combinations to induce osteogenesis or adipogenesis
  • Has been developed and optimized using MSCs
  • Can be used with StemXVivo Osteogenic Supplement (Mfr. Nos. CCM008 or CCM009) or StemXVivo Adipogenic Supplement (Mfr. Nos. CCM011) to reduce variation in differentiation
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Stem Cell Media
250 mL