Learn More
R&D Systems™ StemXVivo Xeno-Free Human MSC Expansion Media
Optimized for the expansion of human mesenchymal stem cells
Brand:  R&D Systems™ CCM021
355.06 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18482350
  • 452.00 € / 500mL
Explore more special offers
Description
Description
Mesenchymal stem cells are often expanded in media that contain serum and other components that are of animal origin. The presence of animal-derived factors in media can introduce experimental variability and limit the use of the cultured cells in translational or clinical studies.
  • Maintains MSCs in an undifferentiated state
  • Free of animal components that can introduce variability and pose a safety risk
  • Suitable for translational studies
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Stem Cell Expansion Media
500 mL