missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Etomoxir sodium salt, Tocris Bioscience™

Product Code. 18752564
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Carnitine palmitoyltransferase I (CPT1) inhibitor

Inhibitor of carnitine palmitoyltransferase I (CPT1); inhibits β-oxidation in mitochondria. Shown to inhibit cardiolipin biosynthesis from exogenous fatty acid in H9c2 cells.

Chemical Identifiers

CAS 828934-41-4
Molecular Formula C15H19ClNaO4
Molecular Weight (g/mol) 321.75
MDL Number MFCD07787411
InChI Key ZUMIFLJZQDMWAU-UHFFFAOYNA-N
Synonym (R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
PubChem CID 131860301
IUPAC Name 2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylic acid sodium
SMILES [Na].OC(=O)C1(CCCCCCOC2=CC=C(Cl)C=C2)CO1

Specifications

Quantity 10 mg
Formula Weight 320.74
Percent Purity >98%
Chemical Name or Material (R)-(+)-Etomoxir sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.