missing translation for 'onlineSavingsMsg'
Learn More
(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 15489475

(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15489475
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15489475

Brand: Thermo Scientific Alfa Aesar H51965.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115962-35-1
Molecular Formula C15H18NO4
Molecular Weight (g/mol) 276.31
MDL Number MFCD00143818
InChI Key HFPVZPNLMJDJFB-GFCCVEGCSA-M
Synonym boc-d-tic-oh, r-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, boc-d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, n-boc-d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3r-2-tert-butoxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r---n-boc-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-2-tert-butoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, r---2-tertbutoxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, boc-d-tic, ambotzbaa1055
PubChem CID 688607
IUPAC Name (3R)-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC2=CC=CC=C2C[C@@H]1C([O-])=O

Specifications

Quantity 5 g
Formula Weight 277.32
Percent Purity 98%
Chemical Name or Material (R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE08Y028 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51965.06,H51965.03
Lot NumberB18W033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51965.06,H51965.03
Lot NumberU30H029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51965.06,H51965.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.