missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-Prolinal, 97+%, Thermo Scientific Chemicals

Product Code. 10550934
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73365-02-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01321389
InChI Key YDBPZCVWPFMBDH-MRVPVSSYSA-N
Synonym n-boc-d-prolinal, n-tert-butoxycarbonyl-d-prolinal, r-tert-butyl 2-formylpyrrolidine-1-carboxylate, boc-d-prolinal, tert-butyl 2r-2-formylpyrrolidine-1-carboxylate, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2r, tert-butyl r-2-formylpyrrolidine-1-carboxylate, r-n-boc-prolinal, pubchem15956
PubChem CID 7009153
IUPAC Name tert-butyl (2R)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C=O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Assay Percent Range 97+%
Infrared Spectrum Authentic
NMR 97.0% min.
Packaging Glass Bottle
Refractive Index 1.4600 to 1.4650 (20°C, 589nm)
Specific Rotation +90° to +102° (20°C, 589nm) (c=1, CHCl3)
Formula Weight 199.25
Density 1.059g/mL
Specific Rotation Condition +91.50° (20°C C=1,chloroform)
Quantity 1 g
Physical Form Liquid
Chemical Name or Material (R)-N-BOC-Prolinal
Show More Show Less
compliance-icons
Product Identifier
  • (R)-N-BOC-Prolinal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'