missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-FMOC-(2-Furyl)alanine, 95%, 98% ee, Thermo Scientific™

Product Code. 10743603 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
This item is not returnable. View return policy

Product Code. 10743603

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 220497-85-8
Molecular Formula C22H19NO5
Molecular Weight (g/mol) 377.40
MDL Number MFCD01311747
InChI Key AJXDCHXGNUFBRC-UHFFFAOYSA-N
Synonym fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc
PubChem CID 16211163
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid
SMILES OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 377.39
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material (R)-N-FMOC-(2-Furyl)alanine, 95%, (98% e.e.)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.