missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-N-Isopropyl-1-phenylethylamine hydrochloride, 98+%, Thermo Scientific™

Product Code. 11374965
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128593-72-6
Molecular Formula C11H18ClN
Molecular Weight (g/mol) 199.722
MDL Number MFCD06011752
InChI Key SVRDLWQPQZOASL-HNCPQSOCSA-N
Synonym r-+-n-isopropyl-1-phenylethylamine hydrochloride, r-n-isopropyl-1-phenylethylamine hydrochloride, isopropyl 1r-1-phenylethyl amine hydrochloride, 1r-1-phenylethyl methylethyl amine, chloride, r-n-2-propyl-1-phenylethylamine hydrochloride, r-n-1-phenylethyl propan-2-amine hydrochloride
PubChem CID 22831491
IUPAC Name N-[(1R)-1-phenylethyl]propan-2-amine;hydrochloride
SMILES CC(C)NC(C)C1=CC=CC=C1.Cl

Specifications

Melting Point 262°C to 265°C
Quantity 1 g
Beilstein 3566584
Sensitivity Hygroscopic
Formula Weight 199.73
Percent Purity ≥98%
Chemical Name or Material (R)-(+)-N-Isopropyl-1-phenylethylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'