Learn More
R&D Systems™ Quantikine Immunoassay Control Group 10
Control for Human High Sensitivity IL-1 beta, IL-4, IL-7, IL-8, IL-10, IL-12 p70, FGF basic, GM-CSF, and TNF-alpha
Brand:  R&D Systems™ QC41
Product Code. 30001352
  • 196.00 € / Each
Explore more special offers
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Control
Human
High Sensitivity IL-1 beta, IL-4, IL-7, IL-8, IL-10, IL-12 p70, FGF basic, GM-CSF, TNF-alpha
1 Each