missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 816 Human Lymphotactin
For use with catalog number DXCL10
Brand: R&D Systems™ QC117
156.80 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
6375 | |
1 Each |
XCL1/Lymphotactin | |
Control | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction