Learn More
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride, Thermo Scientific Chemicals
48.14 € - 490.44 €
Chemical Identifiers
CAS | 138124-32-0 |
---|---|
MDL Number | MFCD02101664 |
InChI Key | YRVXCOIDMFNGIJ-SEILFYAJSA-M |
Synonym | 1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. |
PubChem CID | 131675872 |
IUPAC Name | 2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride |
SMILES | CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10017352
|
Thermo Scientific Acros
295810010 |
1 g |
48.14 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10279273
|
Thermo Scientific Acros
295810050 |
5 g |
135.68 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10195980
|
Thermo Scientific Acros
295810250 |
25 g |
490.44 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
138124-32-0 | |
YRVXCOIDMFNGIJ-SEILFYAJSA-M | |
131675872 | |
CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] |
MFCD02101664 | |
1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. | |
2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride |
Specifications
138124-32-0 | |
100.0 | |
Brown to Yellow | |
Authentic | |
Glass bottle | |
MFCD02101664 | |
1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. | |
CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] | |
635.21 | |
635.21 | |
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride |
97.5 | |
330°C to 332°C | |
96.0 % min. | |
97% | |
C36H52ClMnN2O2 | |
15, 5300 | |
YRVXCOIDMFNGIJ-SEILFYAJSA-M | |
2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride | |
131675872 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO – Research Use Only