missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | RAB5A |
---|---|
Concentration | 0.22 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879744
|
Proteintech
20228-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869744
|
Proteintech
20228-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to RAB5A. It has no cross reaction to RAB5B or RAB5C.
RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.Specifications
RAB5A | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P20339, Q9CQD1 | |
271457, 5868 | |
Peptide | |
Primary | |
-20°C | |
RAB5A |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RAB5, RAB5A, Ras related protein Rab 5A | |
Rab5a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title