missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, Alkaline Phosphatase (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NB7374
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
Affinity Purified | |
RUO | |
Secondary | |
Goat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Western Blot 1:2500 - 1:25000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin | |
Goat IgG-heavy and light chain | |
1 mL | |
Adaptive Immunity, Immunology | |
By immunoelectrophoresis and ELISA this antibody reacts specifically with goat IgG and with light chains common to other goat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Antiserum was cross adsorbed using chicken, horse, human, mouse, pig and rat immunosorbents to remove cross reactive Antibodies. Less than 0.1% cross reactivity to chicken, horse, human, mouse, pig, rabbit and rat IgG was detected. This antibody may cross react with IgG from other species. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction