Learn More
Rabbit anti-Golden Syrian hamster serum proteins, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB9240.2mL
Product Code. 16214640
  • 503.00 € / 2mL
Explore more special offers
Specifications
Rabbit anti-Golden Syrian hamster serum proteins
Polyclonal
Rabbit anti-Golden Syrian hamster IgG recognizes Golden Syrian hamster serum proteins. This secondary antibody was purified using delipidation and defibrination.
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Golden Syrian hamster serum proteins
2 mL
Hamster
Lyophilized
ELISA, Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Rabbit
This product was prepared from polyspecific antiserum by a delipidationand defibrination. Assay by immunoelectrophoresis resulted in a multipleprecipitin arcs against Golden Syrian hamster serum.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.