missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, HRP
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ PA128597
This item is not returnable.
View return policy
Description
PA1-28597 detects Guinea Pig IgG from guinea pig samples.
Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Guinea Pig IgG (H+L) | |
Polyclonal | |
HRP | |
Rabbit | |
IgG fraction | |
RUO | |
Guinea Pig | |
Secondary Antibody | |
IgG |
Immunohistochemistry, ELISA, Western Blot | |
2 mg/mL | |
0.02M potassium phosphate with 1% BSA, 0.15M NaCl and 0.01% gentamicin sulfate; pH 7.2 | |
guinea pig IgG whole molecule. | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
AC4664727D | Certificate of Analysis | 27/03/2025 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, HRP > 1 mg; HRP
Spot an opportunity for improvement?Share a Content Correction