missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG Recombinant Secondary Antibody, Alexa Fluor™ Plus 488, Invitrogen™
Rabbit Recombinant Monoclonal Secondary Antibody
104.00 € - 254.00 €
Specifications
Antigen | Mouse IgG |
---|---|
Clone | 35H35L37 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Western Blot |
Classification | Recombinant Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17896952
|
Invitrogen
A5658050UL |
50 μL |
104.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17857052
|
Invitrogen
A56580 |
250 μL |
254.00 €
250µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgG | |
1 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5 | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
35H35L37 | |
Flow Cytometry, Immunocytochemistry, Western Blot | |
Alexa Fluor Plus 488 | |
Rabbit | |
Mouse | |
IgG | |
Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title