Learn More
Rabbit anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand:  Invitrogen A21210
Product Code. 10368502
  • 253.00 € / Each
Explore more special offers
Description
Description
Product will be shipped at Room Temperature.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgG (H+L) Cross-Adsorbed
Polyclonal
Alexa Fluor 488
Rabbit
purified
RUO
Rat
Liquid
Immunocytochemistry, Immunohistochemistry
2 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains
1 mg
Secondary
4° C, store in dark