Learn More
Shaker, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Shaker.
Brand:  Abnova PAB12776.100ug
Product Code. 16090147
  • 388.00 € / 100µg
Explore more special offers
Specifications
Shaker
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of Shaker.
In TBS, pH 7.2 (BSA, 10% Proclin300)
A synthetic peptide corresponding to N-terminus of Shaker.
RUO
This antibody recognizes ∼73 KDa of Shaker protein.
Store at 4°C. For long term storage store at -20°C to -80°C.
Aliquot to avoid repeated freezing and thawing.
IgG
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Rabbit
100 μg
Primary
Drosophila, Human