Learn More
Rabbit IgG Isotype Control, Invitrogen™
Rabbit Isotype Control
Brand: Invitrogen 31235
217.55 EUR valid until 2024-10-31
Use promo code "21744" to get your promotional price.
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity: Based on immunoelectrophoresis at an antigen concentration of 20 mg/mL, the pattern of precipitation against goat anti-rabbit whole serum is the same as that against goat anti-rabbit IgG, Fc fragment specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Store product at 2-8° C until opened. After opening dilute only enough product for a single day's use. Store remainder at 2-8° C under sterile conditions. Country of Origin: USA
Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.Specifications
Rabbit IgG | |
11.0 mg/mL | |
PBS with no preservative; pH 7.6 | |
Rabbit | |
10 mg | |
Not Applicable | |
Liquid |
Blocking Assay, Control, Immunohistochemistry | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG; Not Applicable | |
purified | |
RUO | |
4° C | |
IgG |