Learn More
Heidolph Instruments Rack For Reax Overhead Shaker
Adaptor for 20 test tubes
Brand:  Heidolph Instruments 549-21000-00
Product Code. 10490063
  • 385.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Allows for simultaneously mixing 20 test tubes o.d. 10 to 18 mm with just one shaker
Specifications
Specifications
Rack
10 to 18 mm
20