Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Brand:  Thermo Scientific™ 1950519
 View more versions of this product
Product Code. 11339291
  • 254.27 € / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • Warranty varies by country, please contact us for details
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Side Access Freezer Rack (Adjustable)
Silver
Stainless Steel
4 Tiers
13.7 cm
23.9 cm
26.8 in.
3 in. Box Racks - Adjustable side access rack
Boxes (3 in.)
15 Boxes/Rack
Thermo Scientific TDE Series and TSX Series (5 shelves)
5.4 in.
9.4 in.
68.1 cm