Learn More
Thermo Scientific™ Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Brand:  Thermo Scientific™ 830030
 View more versions of this product
Product Code. 12938284
  • 290.45 € / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Specialty Rack - 8-Compartment Container
Silver
Square
13 cu. ft. capacity upright freezers
7 in.
5.3 in.
45.7 cm
Specialty Racks - 8-compartment container
General Freezer Items
3 Tiers
18 cm
13.5 cm
18 in.