Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
Shop All Thermo Scientific Products
Click to view available options
Type:
Rack
Rack Insert
Holds:
100-10 mm Tubes
13-25 mm Tubes
20-16 mm Tubes
39-10 mm Tubes
60-16 mm Tubes
8-25 mm Tubes
Depends on insert purchased
Dishes, plates, bags, pouches
For Use With (Equipment):
A25B, A40, S21, S30
A5B,A10B A24B,S49,S19T,S14P,S21P
S13, S12T, S15
Unit Size:
Each
Pack of 2
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
Specifications
| For Use With (Equipment) | A5B,A10B A24B,S49,S19T,S14P,S21P |
| Type | Rack Insert |
| Holds | 100-10 mm Tubes |
| Material | Stainless Steel |