Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
363.81 € - 1346.70 €
Product Code | Brand | Type | For Use With (Equipment) | Holds | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Holds | Price | Quantity & Availability | |||
15425204
|
Thermo Scientific™
185089 |
Canister Rack | All CryoMed controlled-rate freezers | Adjustable, 10-position |
874.35 €
Each |
In Stock
Log in to see stock. |
|||
11736629
|
Thermo Scientific™
4000294 |
Straw Rack | All CryoMed controlled-rate freezers | 40 0.25/0.5 mL Straws |
363.81 €
Each |
In Stock
Log in to see stock. |
|||
11726499
|
Thermo Scientific™
4000303 |
Cane Freezing Rack | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | 162 Canes, 5 to 6 Ampules/Cane |
878.37 €
Each |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||
10265034
|
Thermo Scientific™
4000310 |
Skin Rack | All CryoMed controlled-rate freezers | 48 Skin Packets (2 level) |
1346.70 €
Each |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||
12770348
|
Thermo Scientific™
4000340 |
Canister Rack | All CryoMed controlled-rate freezers | 10 Canisters (for cord blood, Model 4000610) |
792.95 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
11796489
|
Thermo Scientific™
4000700 |
Cane Freezing Rack | All CryoMed controlled-rate freezers | 77 Canes |
1155.75 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||
10633575
|
Thermo Scientific™
4000701 |
Vial Rack (Small) | All CryoMed controlled-rate freezers | 76 1.2 mL/2 mL Ampules |
454.26 €
Each |
In Stock
Log in to see stock. |
|||
10040524
|
Thermo Scientific™
4000702 |
Vial Rack Holder | All CryoMed controlled-rate freezers | Five 1.2 mL/2 mL Cryo Racks (Small) |
557.78 €
Each |
In Stock
Log in to see stock. |
|||
10365954
|
Thermo Scientific™
4000703 |
Vial Rack (Large) | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458, | 161 1.2 mL/2 mL Ampules |
523.61 €
Each |
In Stock
Log in to see stock. |
|||
10244604
|
Thermo Scientific™
4000704 |
Vial Rack Holder | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | Five 1.2 mL/2 mL Cryo Racks (Large) |
611.04 €
Each |
In Stock
Log in to see stock. |
|||
11794898
|
Thermo Scientific™
4000705 |
Vial Rack (Small) | All CryoMed controlled-rate freezers | 76 4 mL/5 mL Ampules |
434.16 €
Each |
In Stock
Log in to see stock. |
|||
11704908
|
Thermo Scientific™
4000706 |
Vial Rack Holder | All CryoMed controlled-rate freezers | Three 4 mL/5 mL Cryo Racks (Small) |
562.80 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
11714908
|
Thermo Scientific™
4000707 |
Vial Rack (Large) | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | 161 4 mL/5 mL Ampules |
508.53 €
Each |
In Stock
Log in to see stock. |
|||
11724908
|
Thermo Scientific™
4000708 |
Vial Rack Holder | CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | Three 4 mL/5 mL Cryo Racks (Large) |
611.04 €
Each |
In Stock
Log in to see stock. |
|||
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details