Learn More
Eppendorf™ RackScan
RackScan reads complete storage racks with up to 96 different barcoded vials. The sample IDs are transferred to downstream databases.
1620.00 €
Specifications
Data Transfer Method PC/USB
Connection Type USB
View More Specs
Products 1
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
16486879
View Documents
Eppendorf™
6002000014
Scanner (Side Reading)
1620.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in accessory
  • Small footprint, ideal for benchtop (and integration)
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Support material of vials available via Eppendorf Homepage for documentation purposes
  • Plug & Transfer with Eppendorf™ eLABInventory and eLABJournal software for comfortable data transfer
  • RackScan s can be attached to RackScan b as accessory to read barcodes on side labels
  • Different formats usable for high flexibility
Specifications
Specifications
PC/USB
USB